Annexin V-FITC Apoptosis Detection Kit with PI, BioLegend
Supplier: BIOLEGEND
640914
10763-240EA 373.1 USD InStock
10763-240
Annexin V-FITC Apoptosis Detection Kit with PI, BioLegend
Assays Cellular Assays
FITC Annexin V Apoptosis Detection Kit with PI is used for the identification of apoptotic and necrotic cells and is suitable for flow cytometric analysis of antibody surface-stained cells.
Annexin V (or Annexin A5) is a member of the annexin family of intracellular proteins that binds to phosphatidylserine (PS) in a calcium dependent manner. Fluorochrome-labeled Annexin V can be used to specifically target and identify apoptotic cells.
Información para pedidos: Cell Staining Buffer is not included and should be ordered separately.
Información de suministro: Kit includes FITC Annexin V 0.5 mL, Propidium Iodide Solution 1 mL and Annexin V Binding Buffer 50 mL.
Precaución: Propidium Iodide Solution is toxigenic and mutagenic; handle with care.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty