Anti-His Tag Mouse Monoclonal Antibody [clone: BHISH8]
Supplier: Biosensis
M-1311-100
10782-048EA 395.85 USD InStock
10782-048
Anti-His Tag Mouse Monoclonal Antibody [clone: BHISH8]
Antibodies
The 6X His tag is a short peptide sequence of 6 histidine residues. Epitopes such as the 6X His tag are often included with the target DNA at the time of cloning to produce fusion proteins containing the tag sequence. This allows anti-epitope tag antibodies such as this one to serve as a universal detection reagent for any recombinant protein containing this tag. Anti-epitope antibodies are a useful alternative to generating antibodies to identify a specific recombinant protein. The 6X His motif is often used as a tag on recombinant proteins to facilitate purification with immobilized metal-More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty