SLIKSTOPPER™ Mat, Brady
Supplier: Brady Worldwide
107768
10809-772RL 590.43 USD InStock
10809-772
SLIKSTOPPER™ Mat, Brady
Spill Control Materials
The SLIKSTOPPER™ is made from chemical-resistant, flexible PVC, this mat seals drains tightly to prevent spills from spreading
Resists water, oil and most aggressive fluids. Easy application and soap and water cleanup for re-use. Vinyl material. Yellow color.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty