Anticon® White Magic™ Cleanroom Wipes, Contec®
Supplier: Contec
495353-303 495353-322 495353-323 495353-317
45800-148CS 683.5 USD InStock
45800-148 10682-794 10682-796 10820-770
Anticon® White Magic™ Cleanroom Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Anticon® White Magic™ Wipes are made from 100% continuous filament polyester with an interlock knit construction. This wipe exhibits low particle and fiber counts and provides ultra-clean wiping for critical applications.
  • Sealed edges for very low levels of particles and fibers
  • Patent manufacturing technology creates as wipe with excellent sorbency
They are ideal for applications that require high sorbency and are excellent for use in electronics applications. These wipes are available validated sterile for life science applications.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty