Ionizer Antistatic Systems, METTLER TOLEDO®
Supplier: Mettler Toledo
63052309 63052494 63052303 63052305 63052306 63052302 63052496 63052495
11238-366EA 1904.28 USD InStock
11238-366 89042-460 11238-356 11238-360 11238-362 11238-354 89042-464 89042-462
Ionizer Antistatic Systems, METTLER TOLEDO®
Ionizers
Antistatic systems discharge electrostatically charged samples for precise weighing.
  • Discharges electrostatically charged samples
  • Neutralizes the disruptive electrostatic charge
  • Eliminates any forces that may distort weighing
A high-voltage, low-current charge neutralizes the disruptive electrostatic charge on the sample, eliminating any forces that may distort weighing.
The systems can be used with any balance and weighing substance. The ionizer instantaneously neutralizes the sample while keeping the weighing pan free and accessible.More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICIOS
Servicios: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty