Draft Chamber Kits, METTLER TOLEDO®
Supplier: Mettler Toledo
63052324 63052337
11277-752EA 585.53 USD InStock
11277-752 11277-985
Draft Chamber Kits, METTLER TOLEDO®
Shields Safety Shields
To help eliminate drafts and stabilize balance readings, these 4.8 mm thick Plexiglas® chambers create a draft-free area around your METTLER TOLEDO® balance.
  • Eliminate drafts
  • Stabilize balance readings
Información para pedidos: Assembly required.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty