VWR® Balance Table
Supplier: VWR International
12567-330
12567-330EA 2362.42 USD InStock
12567-330
VWR® Balance Table
Furniture Tables and Workbenches
Eliminate troublesome vibrations that affect the accuracy of your analytical or microbalances with a VWR® Balance Table.
  • Rigid construction
  • Fast, easy assembly
  • Chemical-resistant finish
This sturdy, engineer-designed table is virtually unshakable
Table is composed of four separate heavy-duty structural elements: two sturdy pedestal bases with non-skid rubber feet, a rigid cross-brace, and a chemical-resistant top.
The table is made using a ¾” high density particle board covered with a high pressure laminated plastic. The center brace is solidMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty