Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Strip Shields, Trax

Supplier: Trax Industrial Products
SS40ST37 SS40ST48 SS40ST68 SS40ST89 SS40ST1010
12999-080EA 455.04 USD InStock
12999-080 12999-082 12999-084 12999-086 12999-088
Strip Shields, Trax
Prefabricated Rooms
Designed for use in applications where access to a clean enclosure is a concern, but traffic patterns are light and restricted to personnel only (no carts or equipment)

Pre-assembled strip shields are 40mil thick and comprise 20.3cm (8") wide material overlapped 5.1cm (2"). Shipping weights vary depending on size.

Información para pedidos: Dual-Lock™ attachment system is included with each strip shield.

Order Now
 

ORDER


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Width Length Supplier No. VWR Catalog Number Unit Price Quantity
3 ft. 7 ft. SS40ST37 12999-080 Each
Retrieving
4 ft. 8 ft. SS40ST48 12999-082 Each
Retrieving
6 ft. 8 ft. SS40ST68 12999-084 Each
Retrieving
8 ft. 9 ft. SS40ST89 12999-086 Each
Retrieving
10 ft. 10 ft. SS40ST1010 12999-088 Each
Retrieving

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty