SP Bel-Art Straining Baskets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products
H16703-0000
13197-334EA 282.14 USD InStock
13197-334
SP Bel-Art Straining Baskets, Bel-Art Products, a part of SP
Baskets Safety Transporting Baskets
Perforated baskets made from polyethylene plastic allow rapid filling and draining, and are handy for batch rinsing and transporting labware as well as straining or filtering large particles.
  • Polyethylene plastic construction
  • Chemically resistant
  • Strap handle rises above the rim for easy handling
Chemically resistant, they withstand acids and alkalis, and have a service temperature up to 82°C (180ºF). Perforations are 2 mm (0.080"), and a strap handle rises 10 cm (4") above the rim.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty