SP Wilmad-LabGlass Church Distilling Flasks, SP Industries
Supplier: Wilmad-LabGlass
LG-7228-100
14202-192EA 184.65 USD InStock
14202-192
SP Wilmad-LabGlass Church Distilling Flasks, SP Industries
Flasks Distillation Flasks
Specified for the distillation of bituminous samples, these borosilicate glass Church Distilling Flasks have a 10 mm I.D
sidearm and a neck made to fit a rubber stopper.
Certificaciones: Reference: ASTM D 20 Test Method for Distillation of Road Tars. ASTM D 246 Method for Distillation of Creosote and Creosote Coal Tar Solutions. ASTM D 402 Test Method for Distillation of Cut-Back Asphaltic (Bituminous) Products. ASTM D 2569 Test Method for Distillation of Pitch.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty