Jenway® Cuvettes for UV and UV-VIS Spectrophotometers, Cole-Parmer
Supplier: Argos Technologies
83070-82
14217-884EA 968.37 USD InStock
14217-884
Jenway® Cuvettes for UV and UV-VIS Spectrophotometers, Cole-Parmer
Cuvettes Spectrophotometer Cuvettes
Cole-Parmer offers an extensive range of cuvettes for measuring liquid samples.
  • Suitable for both UV and visible regions
  • Quartz cuvettes are available in a wide capacity range, from 50 µL to 35ml and pathlengths from 10 to 100 mm
The cuvette required for a specific application will depend on the wavelength range, sample volume available, and pathlength accepted by the spectrophotometer.
Glass cuvettes are only suitable for the visible region, whereas quartz may be used in both the UV and visible region. A 10 mm cuvette typically holds 2 to 3 ml ofMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty