VWR® Specific Gravity Bottle
Supplier: VWR International
16629-009
16629-009EA 319.5 USD InStock
16629-009
VWR® Specific Gravity Bottle
Specific Gravity Bottles
Specific-gravity bottles are flasks made to hold a known volume of liquid at a specified temperature.
  • Borosilicate glass construction
  • Not adjusted
The bottle is weighed, filled with the liquid whose specific gravity is to be found, and weighed again.
Weld type pycnometer devised for specific gravity of volatile liquids. [ST] glass stopper and cap reduce evaporation.
Información para pedidos: Supplied complete with [ST] stopper and [ST] cap.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty