Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Supplier: Gordon Brush
33SSDG-12
22940-584EA 113.55 USD InStock
22940-584
Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Brushes
This brush has 0.003" Stainless Steel Bristle with a Delrin (Acetal) handle.
  • Stainless steel fill material
  • Hand-laced with stainless steel wire into its handle for maximum fiber retention
  • Great for removing stubborn dirt, light rust or corrosion on detailed parts
  • Safely cleans most tarnished metals, but can leave a dull finish on soft metals
Autoclavable for cleanroom, sterile, or medical applications.
Certificaciones: Meets FDA 21 CFR 177.2480 and ASTM D6100 standards.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty