Zinc Plated Steel Handle Applicator Brush, Gordon Brush
Supplier: Gordon Brush
1NG-12
22940-696EA 28.16 USD InStock
22940-696
Zinc Plated Steel Handle Applicator Brush, Gordon Brush
Brushes
An inexpensive brush that is an excellent replacement for disposable acid brushes that quickly shed fibers.
  • Utility duster/cleaner
Zinc plated steel handle holds the fibers in place by 5 tons of mechanical clamping and without any adhesives that could leach out and contaminate surface areas.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty