KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
25055-300 25055-250 25055-500 25055-1000
29114-050CS 775.1 USD InStock
29114-050 89001-264 89001-266 89001-268
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Flasks Round Bottom Flasks
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty