Large Magnetic Demonstration Prism Set
Supplier: United Scientific Supplies
PST620
470002-942EA 204.3 USD InStock
470002-942
Large Magnetic Demonstration Prism Set
Educational Materials Physics Educational Materials Optics Learning Activities
Teach a wide variety of optical principles with this set of seven lenses and prisms
  • Durable storage box
  • Magnetic backing on optical components
An oversized set of 6 lenses and prisms designed for mounting on a magnetic chalk or marker board. Lenses are about 8" long, so your optics experiments would be clearly visible from every part of the classroom. Each piece in the set has a pre-attached magnetic backing. An activity guide is also included. Set comes in a sturdy wooden storage case. Please note: Laser Ray Box not included.
Información paraMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty