Polyethylene Aspirator Filter Pump
Supplier: Bel-Art
F32947-0000_SE
470005-486EA 26.1 USD InStock
470005-486
Polyethylene Aspirator Filter Pump
Pumps Drum/Transfer Pumps
Because fumes and corrosive filter agents do not affect this aspirator, it can be used safely with bases, acids, and strong chemicals. The pump fits any standard laboratory faucet and accepts tubing up to 9.5mm ( 3/8") ID. Operates efficiently on normal water pressure above 11 psi.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty