Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc

Supplier: M.A.Limmer

ZZ0038-1SH
470104-622EA 28 USD InStock
470104-622
Zinc
Chemicals for Science Education
CAS Number: 7440-66-6
Formula: Zn
Density: 7.12 g/mL
Boiling and Freezing Point: 907°C, 419°C
Solubility: Acids and Alkalies
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Información de suministro: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty