Cellular Respiration Kit
Supplier: Edvotek
AP06/285
470121-756EA 89.9 USD InStock
470121-756
Cellular Respiration Kit
Educational Materials Biology Educational Materials AP Biology Learning Activities
For AP Biology Classes
  • Features microrespirometers
  • Time required: ~120 minutes
In this experiment, students learn how to apply the gas laws to the function of the microrespirometer. Students will observe cell respiration of germinating seeds and describe the effects of temperature on the rate of cell respiration.
Kit contents: Pipettes, Vials, Glass beads, Peas, Potassium hydroxide solution (15%), Cork stoppers, Absorbent cotton, Non-absorbent cotton, 1 mL pipettes, Parafilm, Instructions.
Required but not included: trays (10 need to be at leastMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty