Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Parafilm Dispenser

Supplier: Heathrow Scientific
HS234525B
470202-830CS 2491.85 USD InStock
470202-830
Parafilm Dispenser
Sealing Film Dispensers
For maximum protection, this dispenser is designed with a safety razor that slides and cuts with no danger to fingers.

  • ABS Plastic Construction
  • Dispenser Holds One or Two 2" Rolls or one 4" Roll
  • Blue
  • Safety Razor Included

The dispenser holds one or two 2" rolls or one 4" roll.

The dispenser is made of blue ABS plastic.

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty