Vacuum Chamber
Supplier: Thermo Fisher Scientific
5305-0609
470211-850CS 663.3 USD InStock
470211-850
Vacuum Chamber
Vacuum Traps
Nalgene Complete polyetherimide jar and vacuum plate chamber provides strength and good chemical resistance.
Neoprene gasket provides secure seal. Slight amber cast of jar allows complete visibility of interior.
Plate has a center hole for exhaust with tube leading to an external hose connector which accepts 1/4" ID tubing.
For use at room temperature and at full vacuum.
Order Now
ORDER
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Vacuum Chamber, H x OD, in.: 6-5/8 x 9-3/8 5305-0609 470211-850 Each
Retrieving
Case of 2
Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty