Filtration Assembly
Supplier: Wards
FSAS16
470310-690EA 251.7 USD InStock
470310-690
Filtration Assembly
Funnels Filtration Funnels Filtration Apparatus Glass
This filtration assembly is used for vacuum filtration of tissue culture media, biological fluids, HPLC and other aqueous solutions.
  • Unit is designed to maximize flow and reduce foaming
  • Versatile and effective
Assembly consists of 300 ml borosilicate glass funnel, highly integrated sealed 47 mm fritted glass support base, one litre flask. An aluminum clamp is provided to hold the filter funnel and support base properly. The 47 mm filter provides an effective filtration area of 9.6 square centimeters.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty