Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Eisco Brass Test Sieves, 8", Set of 6

Supplier: Wards
BI0139BRSET6
470343-930EA 329.45 USD InStock
470343-930
Eisco Brass Test Sieves, 8", Set of 6
Sieves Test Sieves
Stackable premium quality test sieves for use with many different particle-sizing applications. Brass frames with stainless steel mesh for serial sifting of samples.

  • Features stainless steel woven-wire cloth
  • Sieve is full height 2.5" and measures 203 mm (8") in Ø.

Made of high quality brass and stainless steel - built to last and withstand multiple uses.

Certificaciones: Meets ASTM E11 and AASHTO M 92 requirements for compliance test sieves.

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty