Polypropylene Graduated Cylinders Set
Supplier: Wards
FSC1044
470345-330EA 52.35 USD InStock
470345-330
Polypropylene Graduated Cylinders Set
Cylinders Measuring Cylinders
Set includes eight polypropylene graduated cylinders with hexagonal base, raised graduations, and varying capacities (10 to 2000 ml), offering precise measurement and stability for lab work.
  • Polypropylene graduated cylinders
  • Hexagonal base
  • Autoclavable
  • Perfect classroom or lab starter set
  • Labeled graduations
Polypropylene graduated cylinders, set of 8, hexagonal base, labeled Graduations - 10, 25, 50, 100, 250, 500, 1000, 2000 ml eight cylinders These polypropylene graduated cylinders have a hexagonal base, are made from a single piece and areMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty