Corning® Mouse Collagen IV (from Engelbreth-Holm-Swarm lathrytic mouse tumour)
Supplier: Tianjin Yingte Kemao Corporation
354233
47743-708CS 1299.43 USD InStock
47743-708 47743-733
Corning® Mouse Collagen IV (from Engelbreth-Holm-Swarm lathrytic mouse tumour)
Proteins and Peptides
Collagen IV is a ubiquitous component of basement membranes, the sheet-like matrix that underlies epithelial and endothelial cells and surrounds muscle fat and nerve cells. It can be used as a thin coating on tissue culture surfaces to promote cell attachment and proliferation and to study its effects on cell behaviour.
Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma. Source material tested for hepatitis B antigen and HIV-1 antibody
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty