SP Bel-Art Wet Surface Pen, Bel-Art Products, a part of SP
Supplier: Bel-Art Products
F13382-0000
47750-778EA 51.7 USD InStock
47750-778
SP Bel-Art Wet Surface Pen, Bel-Art Products, a part of SP
Pens Ink Pens
Versatile "space pen" writes clearly on wet, greasy, and other hard to mark surfaces.
  • Compatible for writing on otherwise hard-to-mark surfaces
  • Can be used underwater or in extreme temperatures
  • Writes on dry paper as a standard ballpoint pen
For use on wet labels, in water baths, liquid nitrogen, freezers at –20 °C (–4 °F), on filters while processing underwater-based buffers, for marking slots on Southern, Northern, and Western blots or on vertical surfaces.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty