Digital Wine Refractometer, Model WM-7, ATAGO®
Supplier: Atago
3415-WM-7
47752-868EA 3121.54 USD InStock
47752-868
Digital Wine Refractometer, Model WM-7, ATAGO®
Refractometers Handheld Refractometers
This portable wine refractometer is ideal for winegrowers, wine cooperatives, wineries purchasing grape must, and buyers of grape must
It features seven scales to measure the concentration of grape must accurately and quickly.
Información para pedidos: Supplied with a one-year manufacturer’s warranty. A calibration certificate is available. Contact your Avantor representative for information.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty