SealPlate®, SealPlate® MiniStrips™, and ThinSeal™ Plate Sealing Film, Excel Scientific
Supplier: Excel Scientific
100-SEAL-PLT 100-THIN-PLT SP-2X8-50 SPS-2X8-50 STR-SEAL-PLT STR-THIN-PLT
490007-308BX 104.39 USD InStock
490007-308 490007-312 490007-344 490007-360 490007-362 490007-366
SealPlate®, SealPlate® MiniStrips™, and ThinSeal™ Plate Sealing Film, Excel Scientific
Seals Microplate Seals
SealPlate® 50 µm thickness polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover.
SealPlate® MiniStripsTM sealing films are identical to SealPlate films above, but designed for sealing only one or two 8-well rows at a time on either strip-well or standard plates whenever rows must be selectively protected or accessed. Each MiniStrips film is 19.0 x 103.2 mm, including two positioning tabs. The long dimension with the tabs removed is 82.6 mm. MiniStrips are supplied in sheets of four from which strips canMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty