VWR® Permanent Marking Pens
Supplier: VWR International
52877-864 52877-886 52877-900 52877-810 52877-842
52877-864PK 33.55 USD InStock
52877-864 52877-886 52877-900 52877-810 52877-842
VWR® Permanent Marking Pens
Pens Lab Markers
Perfect marking pens for small lab samples, slides, beakers, sample bags, and more.
  • Available in a wide variety of colors
  • Permanent ink is water- and fade-proof
  • Airtight ring seal cap
Four brilliant colors produce precise lines of 0.3mm width
Write on glass, film, plastic, metal, porcelain, wood, rubber, and paper.
Permanent ink dries in a split second and is water- and fade-proof. Airtight ring seal cap prevents dry-out.
Información para pedidos: Assortment pack consists of six black, two red, two blue, and two green pens.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty