Anti-Static Wire, Justrite®
Supplier: Justrite
08505
56610-593EA 68.82 USD InStock
56610-593
Anti-Static Wire, Justrite®
Wire
Prevents sparks from static electricity generated by movement and flow of flammable liquids.
Bonding between containers during liquid transfer and connecting drums to an earth ground are required under several Federal and Local Codes. Ensure adherence to these codes with this flexible 1 m (3') long insulated wire with 1.5 cm (5/8") C-clamp and 1.5 cm (5/8") alligator clip.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty