VWR® Talon® Chromatography Lab-Frame
Supplier: VWR International
60078-007
60078-007EA 341.36 USD InStock
60078-007
VWR® Talon® Chromatography Lab-Frame
Support Rods and Frames
Specially designed for use with chromatography cabinets.
  • Aluminum construction
  • For use with chromatography cabinets
  • Unique design
Frame features aluminum rods.
Información para pedidos: Lab Frame components include eight 610 mm (24") rods, four rod end connectors, two S-connectors, four Lab Frame feet, and one end-to-end connector.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty