Semi-quantitative test strips QUANTOFIX Nitrite
Supplier: MACHEREY-NAGEL, INC.
91311
60787-734PK 86.75 USD InStock
60787-734
Semi-quantitative test strips QUANTOFIX Nitrite
Environmental Test Kits
For semi-quantitative determinations
Gradation: 0 to 80mg/L. Color changes from white to red/violet.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty