Dial Barometer, Sper Scientific
Supplier: Sper Scientific
736930
61220-786EA 75.05 USD InStock
61220-786
Dial Barometer, Sper Scientific
Barometers
Dial barometer for classroom, lab and industrial use.
Large, clear dual-scale barometer in hPa (hecto Pascal) and mm (millimeter mercury) on a 4" dial which is easy to read from a distance. For classroom, lab, or industrial use. Wall mounts.
Información para pedidos: 5 year warranty.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty