TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
Supplier: Dickson
TH8P3 C415 C417 C480 C472 C473 C476 C477 C481 C478 C479 C456 C436 C482
61279-934EA 1704.83 USD InStock
61279-934 61280-274 61280-276 61280-278 61280-281 61280-282 61280-286 61280-288 61280-290 61280-292 61280-294 61280-298 61280-473 61280-475 61280-284
TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
Chart Recorders Recorders
Units are ideal for remote sensing of temperature and humidity in refrigerators, coolers, cleanrooms, chambers, storage rooms, critical equipment storage, tight locations, and for monitoring IAQ and human comfort
Units 61279-932 and 61279-934 feature 2.5cm (1") 3.5 digit LCD, viewable from 7.6m (25'). Unit 61279-934 features an audio/visual alarm and a remote temperature/relative humidity probe with a 3m (10') cord. All units are portable or wall-mountable.
Recorders have a rugged, compact enclosure with an ABS case, polycarbonate door, and aluminum dial. The large 20.3cm (8") chartMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty