KT8 Thermocouple Remote Sensing Chart Recorders, Dickson®
Supplier: Dickson
KT8P2 KT8P3 C306 C307 C302 C304 C101 C177 C412 C459 C428 C439 C432 C414 C411 C443 C444 C441 C440 C498 C422 C431 C435 C406 C409 C320 C424 C457 C669 C675 C676 C308 C410 C655
61279-922EA 1188.79 USD InStock
61279-922 61279-924 61280-018 61280-022 61280-026 61280-036 61280-042 61280-046 61280-130 61280-224 61280-403 61280-405 61280-407 61280-409 61280-411 61280-413 61280-415 61280-451 61280-453 61280-455 61280-456 61280-468 61280-470 61280-476 61280-478 61280-480 61280-482 61280-485 61280-487 61280-488 61280-504 61280-507 61280-509 61280-510 61280-512 61280-538 61280-544 61280-620 61280-640 61280-644 61280-646 61280-648 61280-650 61280-652 61280-654 61280-656 61280-658 61280-660 61280-662 61280-030 61280-400 61280-563
KT8 Thermocouple Remote Sensing Chart Recorders, Dickson®
Chart Recorders Recorders
These chart recorders do remote monitoring of temperatures in warehouses, food processing facilities, archives, laboratories, and blood storage, dialysis and medical centers.
  • Wall mountable
  • Flip-up pen arm allows for easy chart and pen changes
  • K-thermocouple probe for flexibility, user-selectable temperature ranges and recording times
  • 9V battery backup
Recorders have a rugged, compact enclosure with an ABS case, polycarbonate door, and aluminum dial. The large 20.3 cm (8") charts (sold separately) offer greater resolution. These data acquisitionMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICIOS
Servicios: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty