VWR® Traceable® Alarm Thermometer/Alarm Timer
Supplier: VWR International
62344-916
62344-916EA 104.48 USD InStock
62344-916
VWR® Traceable® Alarm Thermometer/Alarm Timer
Thermometers Digital Thermometers
Unit is ideal for monitoring liquids, air/gas, or semi-solids in water baths, ovens, refrigerators, incubators, and tanks.
  • Traceable®
  • Reads °C and °F
  • Easy-to-read display
  • Chemical-resistant ABS base
Easy-to-read “pop-up” display shows time remaining, probe temperature, and temperature alarm setting
Timer counts down from 23 hours, 59 minutes, 59 seconds in one-minute increments. Alarm sounds when time reaches zero. LCD flashes a times-up message and timer begins counting up. For repetitive applications, memory returns time to previous programmedMore Product Information
Order Now
ORDER
SPECIFICATIONS
SERVICIOS
Servicios: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty