Corning® Hepatocyte Culture Media Kit, Corning
Supplier: Tianjin Yingte Kemao Corporation
355056
62405-898EA 235.75 USD InStock
62405-898
Corning® Hepatocyte Culture Media Kit, Corning
Cell Culture Media
Hepatocyte culture medium is a serum-free, fully-defined medium optimized for hepatocyte differentiation.
Primary human hepatocytes are fresh-couriered in exclusive Falcon® Flip-Lock packaging to ensure best performance and viability upon arrival - without irritating film membranes, gel stabilizers or preservatives that can interfere with your research. Excellent viability- shipped in Corning Hepato-STIM hepatocyte culture medium on Corning® Biocoat™ Multiwell Plates and Flasks for maximal performance. Rapid response- As soon as we have donor tissue, you will be notified instantly as per your instructions.
Información de suministro: Kit comprises hepatocyte culture medium (500 ml) and EGF (5 µg).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty