CONSTIX®, Sealed Foam Swabs, SF-2 Series, Contec®
Supplier: Contec
SF-2
68310-230CS 198.63 USD InStock
68310-230
CONSTIX®, Sealed Foam Swabs, SF-2 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-2 swabs consist of 100ppi sealed closed-cell foam head with a polypropylene handle. The swab head is thermally sealed and bonded to the handle without adhesives or binders.
  • Large rectangular head ideal for swabbing larger areas and applying coatings
The polyurethane foam head provides good particle entrapment and removal. The large size of the foam head of the SF-2 swab provides better surface coverage for cleaning. The SF-2 swab is compatible with many solvents, such as isopropanol and ethanol, but is not recommended for use with ketones,More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty