Blank Paper Disk for AST, Hardy Diagnostics
Supplier: Nabuurs BV
Z7121
75813-972EA 27.44 USD InStock
75813-972
Blank Paper Disk for AST, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
A blank paper disk that can be used for inhibition studies.
  • Prepare your own disks for Kirby Bauer (antimicrobial disk diffusion (AST)) testing
  • Used for inhibition studies
A blank paper disk that can be used for inhibition studies. Used to prepare your own disks for Kirby Bauer (antimicrobial disk diffusion (AST)) testing on Mueller Hinton Agar.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty