Avantor OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, Avantor Fluid Handling
Supplier: Avantor Fluid Handling
15CTAPPCFDSL00 15CTAPPCFDSLST 15CTAPPCFX00 15CTAPPCFXST 15CTAPSCFX00 15CTAPSCFXST 50CTA-CF-DSL00 50CTA-CF-DSLST 50CTA-CF-X-00 50CTA-CF-X-ST
75840-754CS 4771.78 USD InStock
75840-754 75840-756 75840-758 75840-760 75840-764 75840-766 75840-768 75840-770 75840-772 75840-774 75840-776
Avantor OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, Avantor Fluid Handling
Tubes Test Tubes
OmniTop Sample Tubes® are a convenient device that can be used to obtain fluid samples. Each single-use OmniTop tube comes with a pre-attached 0.2 µm vent filter and 18" of tubing (C-Flex®, TYGON®, silicone or PharMed®).
  • Unique cap design allows for complete customization
  • Ability to use different Int.Ø and Ext.Ø tubing diameters
  • Reduced assembly and installation time
  • Eliminate cleaning validation
  • Flexible manufacturing
  • Design permits quick delivery on small lots
  • Closure system can be used with various glass bottles
These tubes are available with orMore Product Information
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty