Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Economical General Purpose Incubators, Yamato

Supplier: Yamato Scientific
IC-913CW IC-403CR IC-413CR IC-603CR IC-613CR IC-803CR IC-813CR IC-903CR IC-913CR IC-103C IC-103CW IC-113CW IC-403CW IC-413CW IC-603CW IC-613CW IC-803CW IC-813CW IC-903CW
75846-600EA 9974.05 USD InStock
75846-600 75846-602 75846-604 75846-606 75846-608 75846-610 75846-612 75846-614 75846-616 75846-702 75846-704 75846-706 75846-708 75846-710 75846-712 75846-714 75846-716 75846-718 75846-720
Economical General Purpose Incubators, Yamato
Incubators Compact Benchtop Incubators
The air-jacket technology of these incubators ensures even and efficient heat distribution throughout the chamber.

  • Natural Convection System
  • Operating Temperature Range: Room Temperature +5 to ~80°C
  • Heater Control Circuit: SSR Drive System
  • Temperature Accuracy: ±1.0 °C
  • Temperature Control Accuracy: ±0.5 °C (at 37 °C)
  • Power Outage Compensation
  • Digital PID Controller

The space-saving incubators have a stainless steel interior and a cold-rolled steel plate exterior with a melamine-resin baking finish

The digital setting display and timer are easy toMore Product Information
Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty