Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methacholine chloride 98, powder_white

Supplier: E. Becker & Co. GmbH
Warning

0219023110 0219023125 0219023180 0219023105
IC19023110EA 80.66 USD InStock
IC19023110 IC19023125 IC19023180 75875-564
Methacholine chloride 98, powder_white
Methacholine chloride
Methacholine chloride is a cholinergic agonist and parasympathomimetic bronchoconstrictor. A metabolically stable analog of acetylcholine. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.
Methacholine is primarily used to diagnose bronchial hyperreactivity, which is the hallmark of asthma and also occurs in chronic obstructive pulmonary disease. Acetyl-β-methylcholine (methacholine), an analog of acetylcholine, is use as a receptor agonist to study processes such as airway hyperresponsiveness associated with asthma.
Metabolically stable analog of acetylcholine; muscarinic agonist. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.

  • Presentation: White Powder
  • Soluble in water (100 mg/mL-clear, colorless solution), alcohol, chloroform; Insoluble in ether.
  • -20°C, desiccate


Formula: C₈H₁₈ClNO₂
MW: 195.69 g/mol
Melting Pt: 164…174 °C
Storage Temperature: Freezer
MDL Number: MFCD00011817
CAS Number: 62-51-1
EINECS: 200-537-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bec30bea35403839184a1bc482dea70cdc3cdab&vid=7bec30bea35403839184a1bc482dea70cdc3cdab&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cf76a0c7a96e5b191eff4fffc1564133bdd5f83&vid=2cf76a0c7a96e5b191eff4fffc1564133bdd5f83&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty