Duran GL45 Cap Membrane for Pressure Applications, Ace Glass Incorporated
Supplier: Ace Glass
7632-08
75877-646PK 81.94 USD InStock
75877-646
Duran GL45 Cap Membrane for Pressure Applications, Ace Glass Incorporated
Closures Bottle Closures
Cap membrane for pressure use is part of the GL45 connection system used with Duran lab bottles.
Cap membrane filter for use in pressure equalization as part of the Duran GL45 Connection System. GL45 Connection Systems are used when a safe transfer of liquid media within a closed and sterile system is required.
Order Now
ORDER
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Pressure Cap Membrane for GL45 Connection System 7632-08 75877-646 Pack of 2
Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty