Intracellular ThiolEZ™ Cell Survival Assay Kit, Rockland Immunochemicals
Supplier: Bandelin Electronic GmbH &Co.KG
KLD-004
75929-182EA 353.13 USD InStock
75929-182
Intracellular ThiolEZ™ Cell Survival Assay Kit, Rockland Immunochemicals
Assays Cellular Assays
Intracellular ThiolEZ™ Cell Survival Assay Kit
  • Intracellular ThiolEZ™ Cell Survival Assay Kit contains enough reagents for 100 assays using a 6-well plate.
Intracellular ThiolEZ™ Cell Survival Assay Kit is an easy to use assay for measurement of thiols in cells; it measures total intracellular thiols including glutathione and cysteine. Glutathione is a natural antioxidant and a major thiol that limits cellular damage in healthy tissues during oxidative stress. It is also involved in repairing damage induced by cancer drugs and radiation and in the detoxification of several commonly used cancer chemotherapeutic drugs. Cysteine, which is a precursor for glutathione synthesis, is a further thiol present in cells. Cysteine is 10 times less abundant than the glutathione. Intracellular thiolEZ™ is ideal for investigators involved in aging research, oxidative stress, antioxidant screening, chemotherapy response and toxicology.
Intracellular ThiolEZ™ Cell Survival Assay Kit contains enough reagents for 100 assays using a 6-well plate.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty