Mouse (Virus Free) Serum Non-Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D223-00
75930-112EA 889.1 USD InStock
75930-112
Mouse (Virus Free) Serum Non-Sterile, Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Mouse Serum is used as a component of bioassays, immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Mouse Serum is also available as a sterile preparationMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty