FreeZone® 4.5L Benchtop Freeze Dryers, Labconco®
Supplier: Labconco
700401040 700401050 700402050 700402070 710401000 710401010 710401070 710402000 720401015 720401030 720401040 720401050 720401070 720402000 700401000 700401010 700401015 700401030 700401070 700402000 700402010 700402015 700402030 700402040 710401015 710401030 710401040 710401050 720402010 720402015 720402030 720402040 720402050 720402070 710402010 710402015 710402030 710402040 710402050 710402070 720401000 720401010
75999-814EA 14836.4 USD InStock
75999-814 75999-816 75999-818 75999-820 75999-822 75999-824 75999-826 75999-828 75999-830 75999-832 75999-834 75999-836 75999-838 75999-840 76000-468 76000-470 76000-472 76000-474 76000-476 76000-478 76000-480 76000-482 76000-484 76000-486 76000-488 76000-490 76000-492 76000-494 76000-496 76000-498 76000-500 76000-502 76000-504 76000-506 76000-636 76000-638 76000-640 76000-642 76000-644 76000-646 76000-648 76000-650
FreeZone® 4.5L Benchtop Freeze Dryers, Labconco®
Freeze Dryers
FreeZone 4.5 Liter Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. Collectors are available to -50° C, -84° C or -105° C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty