Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Type 25A Tandem Cuvette, FireFlySci

Supplier: FIREFLYSCI, INC.
25AUV10
76015-730EA 601.99 USD InStock
76015-730
Type 25A Tandem Cuvette, FireFlySci
Cuvettes Spectrophotometer Cuvettes
Spectrophotometer cuvette divided into two chambers so that users can do two simultaneous measurements.

  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in several different materials: (UV Quartz and IR Quartz)
  • Available in 10 mm lightpath
  • Two windows and partition optically polished
  • Height of partition is approximately 37 mm

Firefly cuvettes are made with high quality quartz and glass material available. GoodMore Product Information
Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty