Solid Closed Screw Cap with PV Lining, FireflySci
Supplier: FIREFLYSCI, INC.
SC1W
76016-672EA 19.4 USD InStock
76016-672
Solid Closed Screw Cap with PV Lining, FireflySci
Closures Bottle Closures
Closed screw cap comes with PTFE septa.
  • Caps are made of polypropylene with poly-vinyl liner
  • Good for mild acids, alkalis, solvents, alcohols, oils, and aqueous products
Caps may be used with a broad range of FireflySci cuvettes.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty