proANP ELISA kit, Enzo Life Sciences
KIT153-0001
76045-580EA 736 USD InStock
76045-580
proANP ELISA kit, Enzo Life Sciences
Assays ELISAs
Sensitive proANP ELISA kit enabling detection of proANP in serum, plasma, urine and cell culture supernatant in just 3.5 hours.
For the quantitative determination of human proANP (1-98) in serum, plasma, urine, and cell culture supernatants.
The proANP ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3.5 hours.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty