Guilford-Wright Wire Cutting Scissors, OR Grade, Sklar®
Supplier: Sklar
24-2350
76192-976EA 276.56 USD InStock
76192-976
Guilford-Wright Wire Cutting Scissors, OR Grade, Sklar®
Scissors
Sklar® toothed Guilford-Wright suture wire cutting scissors are non-ratcheted, finger ring instruments used for cutting sutures in preparation for removal.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Blades are much shorter than the shanks. The overall length is shorter than the Smith.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty